3-Methoxy-2,2-dimethylpropan-1-ol 95%

Lieferant: Apollo Scientific

OR916644-1G
APOSOR916644-1GEA 502 EUR
APOSOR916644-1G
3-Methoxy-2,2-dimethylpropan-1-ol 95%
3-Methoxy-2,2-dimethylpropan-1-ol
Formel: C₆H₁₄O₂
Molecular Weight: 118,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09908260
CAS-Nummer: 57021-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR