3-Bromo-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxylic acid 96%

Lieferant: Apollo Scientific

OR916671-25G
APOSOR916671-25GEA 68.3 EUR
APOSOR916671-25G
3-Bromo-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxylic acid 96%
3-Bromo-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxylic acid
Formel: C₉H₅BrClN₃O₂
Molecular Weight: 302.512 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689880
CAS-Nummer: 500011-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR