5-Phenyl-pyrimidine 95%

Lieferant: Apollo Scientific

OR916707-1G
APOSOR916707-1GEA 806 EUR
APOSOR916707-1G
5-Phenyl-pyrimidine 95%
5-Phenyl-pyrimidine
Formel: C₁₀H₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00234616
CAS-Nummer: 34771-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR