(R)-(+)-2-Bromisovaleriansäure 95%

Lieferant: Apollo Scientific

Synonyme: (R)-(+)-2-Bromo-3-methylbutyric acid, (R)-2-Bromoisovaleric acid, (R)-2-Bromo-3-methylbutyric acid

OR916712-5G
APOSOR916712-5GEA 139 EUR
APOSOR916712-5G
(R)-(+)-2-Bromisovaleriansäure 95%
(R)-(+)-2-Bromisovaleriansäure
Formel: C₅H₉BrO₂
Molecular Weight: 181,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210113
CAS-Nummer: 76792-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR