Methyl 4,5-Dibromo-2-furancarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR916713-1G
OR916713-5G
APOSOR916713-1GEA
66.7
EUR
APOSOR916713-1G
APOSOR916713-5G
Methyl 4,5-Dibromo-2-furancarboxylate
Methyl 4,5-Dibromo-2-furancarboxylate
Formel:
C₆H₄Br₂O₃ Schmelzpunkt: 56…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092493 CAS-Nummer: 54113-41-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...