4,6-Dimethoxypyrimidin-5-amine 95%

Lieferant: Apollo Scientific

OR916730-1G
APOSOR916730-1GEA 79.9 EUR
APOSOR916730-1G
4,6-Dimethoxypyrimidin-5-amine 95%
4,6-Dimethoxypyrimidin-5-amine
Formel: C₆H₉N₃O₂
Molecular Weight: 155,1546 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00194013
CAS-Nummer: 15846-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR