(Phenylthio)essigsäure 95%
Lieferant: Apollo Scientific
OR916733-100G
APOSOR916733-100GEA
163
EUR
APOSOR916733-100G
(Phenylthio)essigsäure 95%
(Phenylthio)essigsäure
Formel:
C₈H₈O₂S Molecular Weight: 168,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004355 CAS-Nummer: 103-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...