3-(2-Cyanophenyl)propanoic acid 95%

Lieferant: Apollo Scientific

OR916736-1G
APOSOR916736-1GEA 151 EUR
APOSOR916736-1G
3-(2-Cyanophenyl)propanoic acid 95%
3-(2-Cyanophenyl)propanoic acid
Formel: C₁₀H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00464857
CAS-Nummer: 27916-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR