2-(4-Methoxycyclohexyl)ethanamin (cis/trans-Gemisch) 95%

Lieferant: Apollo Scientific

OR916743-1G
APOSOR916743-1GEA 538 EUR
APOSOR916743-1G
2-(4-Methoxycyclohexyl)ethanamin (cis/trans-Gemisch) 95%
2-(4-Methoxycyclohexyl)ethylamine (cis- and trans- mixture)
Formel: C₉H₁₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07772981
CAS-Nummer: 1052223-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR