7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline 95%

Lieferant: Apollo Scientific

OR916749-1G OR916749-5G
APOSOR916749-1GEA 0 EUR
APOSOR916749-1G APOSOR916749-5G
7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline 95%
7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline
Formel: C₁₀H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00143517
CAS-Nummer: 4602-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR