3-Ethyl-2-methyl-3-pentanol 95%
Lieferant: Apollo Scientific
Synonyme:
1,1-Diethylisobutyl Alcohol, Diethylisopropylcarbinol
OR916791-5G
APOSOR916791-5GEA
415
EUR
APOSOR916791-5G
3-Ethyl-2-methyl-3-pentanol 95%
3-Ethyl-2-methyl-3-pentanol
Formel:
C₈H₁₈O Molecular Weight: 130,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046589 CAS-Nummer: 597-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...