1,2-Indanedione 97%
Lieferant: Apollo Scientific
OR916829-10G
OR916829-5G
OR916829-25G
APOSOR916829-10GEA
477
EUR
APOSOR916829-10G
APOSOR916829-5G
APOSOR916829-25G
1,2-Indanedione 97%
1,2-Indanedione
Formel:
C₉H₆O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463378 CAS-Nummer: 16214-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...