L-4-Methoxymandelic acid 95%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Hydroxy-2-(4-methoxyphenyl)acetic acid

OR916843-1G
APOSOR916843-1GEA 72.5 EUR
APOSOR916843-1G
L-4-Methoxymandelic acid 95%
L-4-Methoxymandelic acid
Formel: C₉H₁₀O₄
Molecular Weight: 182,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00754498
CAS-Nummer: 75172-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR