Quinoxalin-6-ylmethanol 95%
Lieferant: Apollo Scientific
OR916865-1G
APOSOR916865-1GEA
290
EUR
APOSOR916865-1G
Quinoxalin-6-ylmethanol 95%
Quinoxalin-6-ylmethanol
Formel:
C₉H₈N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06227444 CAS-Nummer: 488834-75-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...