5-Bromo-2-chloro-4-hydroxypyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-chloro-pyridin-4-ol
OR916894-250MG
APOSOR916894-250MGEA
39.3
EUR
APOSOR916894-250MG
5-Bromo-2-chloro-4-hydroxypyridine 95%
5-Bromo-2-chloro-4-hydroxypyridine
Formel:
C₅H₃BrClNO Molecular Weight: 208,44 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189877 CAS-Nummer: 1196146-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...