4-Chlor-2-isopropyl-5-methylphenol 98%

Lieferant: Apollo Scientific

Synonyme: 6-Chlorthymol

OR916910-25G
APOSOR916910-25GEA 127 EUR
APOSOR916910-25G
4-Chlor-2-isopropyl-5-methylphenol 98%
4-Chlor-2-isopropyl-5-methylphenol
Formel: C₁₀H₁₃ClO
Molecular Weight: 184.67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002326
CAS-Nummer: 89-68-9
Merck Index: 13,02189

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR