Ethyl 1-ethylpyrazole-4-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 1-Ethylpyrazole-4-carboxylate

OR916923-1G
APOSOR916923-1GEA 169 EUR
APOSOR916923-1G
Ethyl 1-ethylpyrazole-4-carboxylate 95%
Ethyl 1-ethylpyrazole-4-carboxylate
Formel: C₈H₁₂N₂O₂
Molecular Weight: 168,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16620009
CAS-Nummer: 447401-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR