Iproniazid 95%
Lieferant: Apollo Scientific
Synonyme:
Isonicotinic acid 2-Isopropylhydrazide, 1-Isonicotinoyl-2-isopropylhydrazine
OR916950-1G
OR916950-250MG
APOSOR916950-1GEA
88.6
EUR
APOSOR916950-1G
APOSOR916950-250MG
Iproniazid 95%
Iproniazid
Formel:
C₉H₁₃N₃O Molecular Weight: 179.22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153819 CAS-Nummer: 54-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...