Cyclohexylsulphamoyl chloride 95%

Lieferant: Apollo Scientific

OR917027-1G OR917027-250MG
APOSOR917027-1GEA 0 EUR
APOSOR917027-1G APOSOR917027-250MG
Cyclohexylsulphamoyl chloride 95%
Cyclohexylsulphamoyl chloride
Formel: C₆H₁₂ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11109158
CAS-Nummer: 10314-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR