N-Cyclopropyl-1H-imidazole-1-carboxamide 95%

Lieferant: Apollo Scientific

OR917028-1G
APOSOR917028-1GEA 435 EUR
APOSOR917028-1G
N-Cyclopropyl-1H-imidazole-1-carboxamide 95%
N-Cyclopropyl-1H-imidazole-1-carboxamide
Formel: C₇H₉N₃O
Molecular Weight: 151,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11099699
CAS-Nummer: 1033090-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR