Methoxycyclohexan 95%
Lieferant: Apollo Scientific
Synonyme:
Cyclohexyl methyl ether
OR917041-5G
APOSOR917041-5GEA
415
EUR
APOSOR917041-5G
Methoxycyclohexan 95%
Methoxycyclohexane
Formel:
C₇H₁₄O Molecular Weight: 114,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00154087 CAS-Nummer: 931-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...