4-Isopropyl-m-kresol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Isopropyl-3-methylphenol, 4-Isopropyl-3-hydroxytoluol
OR917093-100G
APOSOR917093-100GEA
46.4
EUR
APOSOR917093-100G
4-Isopropyl-m-kresol 95%
4-Isopropyl-m-kresol
Formel:
C₁₀H₁₄O Molecular Weight: 150,22 g/mol Schmelzpunkt: 111…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010704 CAS-Nummer: 3228-02-2 EINECS: 221-761-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...