(2-phenoxyphenyl)methanol 97%

Lieferant: Apollo Scientific
Achtung

OR9171-100MG
APOSOR9171-100MGEA 0 EUR
APOSOR9171-100MG
(2-phenoxyphenyl)methanol 97%
(2-phenoxyphenyl)methanol
Formel: C₁₃H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00017297
CAS-Nummer: 13807-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR