Cis-7-azabicyclo[3.3.0]octane 95%
Lieferant: Apollo Scientific
OR917123-1G
APOSOR917123-1GEA
348
EUR
APOSOR917123-1G
Cis-7-azabicyclo[3.3.0]octane 95%
Cis-7-azabicyclo[3.3.0]octane
Formel:
C₇H₁₃N Molecular Weight: 111,185 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953171 CAS-Nummer: 926276-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...