2-(4-Chlorphenyl)-2-methyl-1-propanol 98%
Lieferant: Apollo Scientific
OR917141-5G
APOSOR917141-5GEA
219
EUR
APOSOR917141-5G
2-(4-Chlorphenyl)-2-methyl-1-propanol 98%
2-(4-Chlorophenyl)-2-methylpropanol
Formel:
C₁₀H₁₃ClO Molecular Weight: 184,66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657569 CAS-Nummer: 80854-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...