Pyrimidin-4-ol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR917181-25G
APOSOR917181-25GEA
121
EUR
APOSOR917181-25G
Pyrimidin-4-ol 98%
Pyrimidin-4-ol
Formel:
C4H4N2O Schmelzpunkt: 163…169 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00038024 CAS-Nummer: 51953-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...