[4-(Pyridin-2-yl)phenyl]methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR9172-1G
APOSOR9172-1GEA
318
EUR
APOSOR9172-1G
[4-(Pyridin-2-yl)phenyl]methanol 97%
[4-(Pyridin-2-yl)phenyl]methanol
Formel:
C₁₂H₁₁NO Schmelzpunkt: 49…51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116292 CAS-Nummer: 98061-39-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...