3,4-Dichloro-5-nitropyridine 95%

Lieferant: Apollo Scientific

OR917240-1G
APOSOR917240-1GEA 52.2 EUR
APOSOR917240-1G
3,4-Dichloro-5-nitropyridine 95%
3,4-Dichloro-5-nitropyridine
Formel: C₅H₂Cl₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09910258
CAS-Nummer: 56809-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR