N-(Carbobenzoxy)aminoacetonitrile 95%

Lieferant: Apollo Scientific

Synonyme: Benzyl (cyanomethyl)carbamate, Cyanomethylcarbamic Acid Benzyl Ester, Benzyl Cyanomethylcarbamate, N-Cbz-aminoacetonitrile

OR917260-25G
APOSOR917260-25GEA 237 EUR
APOSOR917260-25G
N-(Carbobenzoxy)aminoacetonitrile 95%
N-(Carbobenzoxy)aminoacetonitrile
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00033175
CAS-Nummer: 3589-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR