sec-Butyl chloroformate 95%

Lieferant: Apollo Scientific

OR917261-1G OR917261-25G OR917261-5G
APOSOR917261-1GEA 0 EUR
APOSOR917261-1G APOSOR917261-25G APOSOR917261-5G
sec-Butyl chloroformate 95%
sec-Butyl chloroformate
Formel: C₅H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00043796
CAS-Nummer: 17462-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR