Piperidin-4-yl-acetic acid tert-butyl ester 95%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-(piperidin-4-yl)acetate, tert-butyl 2-(4-piperidyl)acetate

OR917271-1G
APOSOR917271-1GEA 223 EUR
APOSOR917271-1G
Piperidin-4-yl-acetic acid tert-butyl ester 95%
Piperidin-4-yl-acetic acid tert-butyl ester
Formel: C₁₁H₂₁NO₂
Molecular Weight: 199,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738957
CAS-Nummer: 180182-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR