(4-Bromo-1-methyl-1H-pyrazol-5-yl)methanol 95%
Lieferant: Apollo Scientific
OR917350-1G
APOSOR917350-1GEA
339
EUR
APOSOR917350-1G
(4-Bromo-1-methyl-1H-pyrazol-5-yl)methanol 95%
(4-Bromo-1-methyl-1H-pyrazol-5-yl)methanol
Formel:
C₅H₇BrN₂O Molecular Weight: 191,026 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18793417 CAS-Nummer: 1276056-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...