trans-Decahydroquinoline 98%
Lieferant: Apollo Scientific
OR917395-100G
OR917395-25G
OR917395-5G
APOSOR917395-100GEA
806
EUR
APOSOR917395-100G
APOSOR917395-25G
APOSOR917395-5G
trans-Decahydroquinoline 98%
trans-Decahydroquinoline
Formel:
C₉H₁₇N Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137755 CAS-Nummer: 767-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...