trans-Decahydroquinoline 98%

Lieferant: Apollo Scientific

OR917395-100G OR917395-25G OR917395-5G
APOSOR917395-100GEA 806 EUR
APOSOR917395-100G APOSOR917395-25G APOSOR917395-5G
trans-Decahydroquinoline 98%
trans-Decahydroquinoline
Formel: C₉H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD00137755
CAS-Nummer: 767-92-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR