[(2,6-Dimethylphenyl)amino](oxo)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dimethylanilino(oxo)acetic acid, 2-((2,6-Dimethylphenyl)amino)-2-oxoacetic acid, N-(2,6-Dimethylphenyl)oxamic acid, DMPAO, (2,6-Dimethylanilino)(oxo)acetic acid, [(2,6-Dimethylphenyl)Carbamoyl]formic acid

OR917428-5G
APOSOR917428-5GEA 101 EUR
APOSOR917428-5G
[(2,6-Dimethylphenyl)amino](oxo)essigsäure 98%
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193.20 g/mol
Schmelzpunkt: 173 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD10018453
CAS-Nummer: 2903-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR