4-Methyl-1-phenyl-2-pentanol 96%
Lieferant: Apollo Scientific
OR917429-25G
APOSOR917429-25GEA
115
EUR
APOSOR917429-25G
4-Methyl-1-phenyl-2-pentanol 96%
4-Methyl-1-phenyl-2-pentanol
Formel:
C₁₂H₁₈O Molecular Weight: 178.28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209515 CAS-Nummer: 7779-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...