Ac-4,5-dehydro-Leu-oh 95%

Lieferant: Apollo Scientific

OR917451-250MG
APOSOR917451-250MGEA 502 EUR
APOSOR917451-250MG
Ac-4,5-dehydro-Leu-oh 95%
Ac-4,5-dehydro-Leu-oh
Formel: C₈H₁₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00237465
CAS-Nummer: 88547-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR