8-Chloroquinoline-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR917481-1G
APOSOR917481-1GEA 148 EUR
APOSOR917481-1G
8-Chloroquinoline-2-carbaldehyde 95%
8-Chloroquinoline-2-carbaldehyde
Formel: C₁₀H₆ClNO
Molecular Weight: 191,614 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06407641
CAS-Nummer: 59394-28-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR