3-Phenoxypropan-1-ol 95%

Lieferant: Apollo Scientific

Synonyme: 3-Phenoxy-1-propanol

OR917493-100G
APOSOR917493-100GEA 132 EUR
APOSOR917493-100G
3-Phenoxypropan-1-ol 95%
3-Phenoxypropan-1-ol
Formel: C₉H₁₂O₂
Molecular Weight: 152,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039549
CAS-Nummer: 6180-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR