6-Methyl-1H-pyrazolo[3,4-b]pyridine 95%

Lieferant: Apollo Scientific

OR917539-1G
APOSOR917539-1GEA 200 EUR
APOSOR917539-1G
6-Methyl-1H-pyrazolo[3,4-b]pyridine 95%
6-Methyl-1H-pyrazolo[3,4-b]pyridine
Formel: C₇H₇N₃
Molecular Weight: 133,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234559
CAS-Nummer: 885269-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR