(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(-)-2,3:4,6-Di-O-isopropylidene-alpha-L-xylo-2-hexulofuranosonic acid monohydrate, 2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonicacidmonohydrate, 2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate, Dikegulac monohydrate
OR917577-25G
APOSOR917577-25GEA
29
EUR
APOSOR917577-25G
(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate 98%
(-)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate
Formel:
C₁₂H₁₈O₇·H₂O Molecular Weight: 274,27 g/mol Schmelzpunkt: 88 - 90 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00150517 CAS-Nummer: 68539-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...