N-Acetyl-DL-Norleucin 95%

Lieferant: Apollo Scientific

OR917582-5G
APOSOR917582-5GEA 0 EUR
APOSOR917582-5G
N-Acetyl-DL-Norleucin 95%
N-Acetyl-DL-Norleucin
Formel: C₈H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00037272
CAS-Nummer: 7682-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR