3-Oxa-9-azaspiro[5.5]undecane

Lieferant: Apollo Scientific
Gefahr

OR917623-250MG
APOSOR917623-250MGEA 169 EUR
APOSOR917623-250MG
3-Oxa-9-azaspiro[5.5]undecane
3-Oxa-9-azaspiro[5.5]undecane
Formel: C₉H₁₇NO
MDL: MFCD11505644
CAS-Nummer: 311-21-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR