exo-Norborneol 98%

Lieferant: Apollo Scientific

OR917646-5G
APOSOR917646-5GEA 71.2 EUR
APOSOR917646-5G
exo-Norborneol 98%
exo-Norborneol
Formel: C₇H₁₂O
Molecular Weight: 112,17 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 497-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR