N-Isopropyl-1H-imidazole-1-carboxamide 95%

Lieferant: Apollo Scientific

OR917725-1G
APOSOR917725-1GEA 464 EUR
APOSOR917725-1G
N-Isopropyl-1H-imidazole-1-carboxamide 95%
N-Isopropyl-1H-imidazole-1-carboxamide
Formel: C₇H₁₁N₃O
Molecular Weight: 153,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11099729
CAS-Nummer: 75472-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR