2,3-Dihydroxybenzonitril 95%

Lieferant: Apollo Scientific

OR917737-5G
APOSOR917737-5GEA 102 EUR
APOSOR917737-5G
2,3-Dihydroxybenzonitril 95%
2,3-Dihydroxybenzonitril
Formel: C₇H₅NO₂
Molecular Weight: 135.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02261930
CAS-Nummer: 67984-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR