Methyl 3-[(4-hydroxyphenyl)amino]propanoate 95%
Lieferant: Apollo Scientific
OR917745-1G
APOSOR917745-1GEA
290
EUR
APOSOR917745-1G
Methyl 3-[(4-hydroxyphenyl)amino]propanoate 95%
Methyl 3-[(4-hydroxyphenyl)amino]propanoate
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195,215 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20666958 CAS-Nummer: 70156-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...