tert-Butyl crotonate 95%

Lieferant: Apollo Scientific

OR917764-25G OR917764-5G
APOSOR917764-25GEA 371 EUR
APOSOR917764-25G APOSOR917764-5G
tert-Butyl crotonate 95%
tert-Butyl crotonate
Formel: C₈H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00059055
CAS-Nummer: 79218-15-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR