1-Methoxy-2-methylpropan-2-amine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: (2-Methoxy-1,1-dimethylethyl)amine Hydrochloride

OR917849-1G
APOSOR917849-1GEA 382 EUR
APOSOR917849-1G
1-Methoxy-2-methylpropan-2-amine hydrochloride 95%
1-Methoxy-2-methylpropan-2-amine hydrochloride
Formel: C₅H₁₃NO·ClH
Molecular Weight: 139,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16036163
CAS-Nummer: 251660-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR