2-Amino-5-nitrobenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Amino-5-nitrophenyl)methanol

OR917871-1G
APOSOR917871-1GEA 65.4 EUR
APOSOR917871-1G
2-Amino-5-nitrobenzyl alcohol 97%
2-Amino-5-nitrobenzyl alcohol
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
Schmelzpunkt: 130…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09834918
CAS-Nummer: 77242-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR